missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H3.1, Monomethyl Arg128 Antibody (4G12), Novus Biologicals™
Rabbit Monoclonal Antibody
343.00€ - 533.00€
Specifications
| Antigen | Histone H3.1 |
|---|---|
| Clone | 4G12 |
| Dilution | Western Blot 1:500-1:2000, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30030594
|
Novus Biologicals
NBP3-26132-50ul |
50 μL |
343.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30028130
|
Novus Biologicals
NBP3-26132-100ul |
100 μL |
533.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Histone H3.1 Monoclonal antibody specifically detects Histone H3.1 in Human, Mouse samples. It is validated for Western Blot, ELISASpecifications
| Histone H3.1 | |
| Western Blot 1:500-1:2000, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| H3 histone family, member D, H3.1HIST1H3D, H3/d, H3FA, H3FB, H3FC, H3FDHIST1H3F, H3FF, H3FH, H3FI, H3FJ, H3FK, H3FL, HIST1H3A, HIST1H3B, HIST1H3C, HIST1H3G, HIST1H3H, HIST1H3I, HIST1H3J, histone 1, H3e, histone cluster 1, H3e, histone H3.1, Histone H3/a, Histone H3/b, Histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h, Histone H3/i, Histone H3/j, Histone H3/k, Histone H3/l | |
| Synthesized peptide [UniProt P68431] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 4G12 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Epigenetics, Phospho Specific | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 8353 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title