missing translation for 'onlineSavingsMsg'
Learn More

Histone H3.3, p Ser31 Antibody (SR1217), Novus Biologicals™

Rabbit Monoclonal Antibody

338.00€ - 449.00€

Specifications

Antigen Histone H3.3
Clone SR1217
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18650725
View Documents
Novus Biologicals
NBP3-21599-100ul
100 μg
449.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18671014
View Documents
Novus Biologicals
NBP3-21599-50ul
50 μg
338.00€
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Histone H3.3 Monoclonal antibody specifically detects Histone H3.3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

Histone H3.3
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200
Monoclonal
Purified
RUO
Human, Mouse, Rat
H3.3A, H3F3, H3F3A, Histone H3.3, Histone H3.3A, Histone H3.3B, MGC87782, MGC87783
A synthesized peptide derived from human phospho-Histone H3.3 [Ser31] (Uniprot #: P84243)
Primary
Store at -20°C. Avoid freeze/thaw cycles.
SR1217
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
Unconjugated
Rabbit
Chromatin Research, Epigenetics
PBS (pH 7.4), 150 mM NaCl, 50% Glycerol
3020
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.