Learn More
Histone H3 [Monomethyl Lys56] Antibody (RM180) [Janelia Fluor™ 669], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-26023JF669
Product Code. 30167973
  • 790.00€ / 0.10mL
Description
Histone H3 Monoclonal antibody specifically detects Histone H3 in Human,Vertebrate samples. It is validated for ELISA,Western Blot,Multiplex Immunoassay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Histone H3
Monoclonal
Janelia Fluor 669
H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783
This Histone H3 [Monomethyl Lys56] antibody (RM180) was raised against a monomethyl-peptide corresponding to Histone H3 [Monomethyl Lys56]
0.1 mL
Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls
126961
Store at 4°C in the dark.
IgG
ELISA, Western Blot, Multiplex
RM180
50mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Human, Vertebrates