Specifications
| Column Format | HiTrap™ |
| For Use With | Purification of IgM |
| Quantity | 1 Ea. |
| Volume (Metric) | 5 mL |
| Type | IgM Purification Column |
| Technique | Antibody Affinity Chromatography |
Safety and Handling
Signal Word
- Warning
- Flammable liquid Category 3