Learn More
HJ 592-2010 Nitroaromatics Mixture 543 100 μg/mL in Methanol, Dr. Ehrenstorfer
Product Code. 30324134
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30324134 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30324134 Supplier Dr. Ehrenstorfer Supplier No. DREA50000543ME
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
CAS 99-35-4, 99-08-1, 118-96-7, 121-14-2, 606-20-2, 88-72-2
Quantity 1 mL
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol