missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HLA-ABC Monoclonal Antibody (W6/32), NovaFluor™ Yellow 660, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
178.00€ - 392.00€
Specifications
| Antigen | HLA-ABC |
|---|---|
| Clone | W6/32 |
| Concentration | 0.6 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055211
|
Invitrogen™
H020T03Y04-A |
100 Tests |
392.00€
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30055147
|
Invitrogen™
H020T02Y04-A |
25 Tests |
178.00€
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HLA-ABC Monoclonal antibody specifically detects HLA-ABC in Human samples. It is validated for Flow CytometrySpecifications
| HLA-ABC | |
| 0.6 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human | |
| P01889, P04439, P10321 | |
| 3105, 3106, 3107 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| W6/32 | |
| Flow Cytometry | |
| NovaFluor Yellow 660 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| A29 protein; alpha chain of MHC class II; antigen presenting molecule; AS; Aw-33; Aw-74; B-4901; B-5001; Bw-47; Bw-50; class I MHC, HLA-B antigen; D6S204; epididymis secretory protein Li 83; H-2K; H2-K; H-2K(d); HEL-S-83; histocompatibility antigen; HLA A; HLA -B; HLA B-40011; HLA B44; HLA C; HLA class I antigen; HLA class I antigen HLA-B; HLA class I histocompatibility antigen C alpha chain; HLA class I histocompatibility antigen Cw-1 alpha chain; HLA class I histocompatibility antigen, A alpha chain; HLA class I histocompatibility antigen, A-1 alpha chain; HLA class I histocompatibility antigen, A-11 alpha chain-like; HLA class I histocompatibility antigen, A-30 alpha chain; HLA class I histocompatibility antigen, A-33 alpha chain; HLA class I histocompatibility antigen, A-74 alpha chain; HLA class I histocompatibility antigen, B alpha chain; HLA class I histocompatibility antigen, B-47 alpha chain; HLA class I histocompatibility antigen, B-50 alpha chain; HLA class I histocompatibility antigen, B-82 alpha chain; HLA class I histocompatibility antigen, C alpha chain; HLA class I histocompatibility antigen, Cw-1 alpha chain; HLA class I histocompatibility antigen, Cw-15 alpha chain; HLA class I histocompatibility antigen, Cw-18 alpha chain; HLA locus; HLA molecule; HLAA; HLA-A; HLA-A locus alpha 2 domain; HLA-A null; HLA-A11; HLA-A33; HLA-Aw33.1; HLAB; HLA-B; HLA-B 1502; HLA-B alpha chain; HLA-B alpha chain (B*2706); HLA-B alpha chain (B*5002); HLA-B alpha-chain; HLA-B*45ZJ; HLA-B15; HLA-B35; HLA-B35 antigen; HLA-B-3506; HLA-B39; HLA-B-3905; HLA-B49; HLA-B50; HLA-B55; HLA-B-5502; HLA-B-5602; HLA-B59; HLA-B61; HLA-Bw50 antigen; HLAC; HLA-C; HLA-C alpha chain; HLA-C alpha chain (HLA-Cw*15v); HLA-C alpha chain for Cw*18GB; HLA-C alpha-chain; HLA-C antigen; HLA-Cw; HLA-Cw*04GB; HLA-Cw*1702; HLA-Cw*18GB; HLA-Cw-1503; HLA-DQB1; HLA-DRB1; HLA-JY3; HLC-C; Human leukocyte antigen A; Human leukocyte antigen C; human leukocyte antigen-C alpha chain; hypothetical protein; lecocyte antigen B; leucocyte antigen A; Leukocyte Antigen; leukocyte antigen C; leukocyte antigen class I-A; leukocyte antigen class I-B; Leukoocyte Antigen; LOC100515902; LOW QUALITY PROTEIN: HLA class I histocompatibility antigen, A-11 alpha chain-like; lymphocyte antigen; major histocompatability complex B antigen; major histocompatibility antigen HLA-C; major histocompatibility complex, class I, A; major histocompatibility complex, class I, B; major histocompatibility complex, class I, C; MCH class I antigen; MGC184092; MGC7052; MHC; MHC 1; MHC antigen; MHC clas I antigen; MHC Class 1; MHC class 1 antigen; MHC class I; MHC class I alpha chain; MHC class I anitgen; MHC class I anti; MHC Class I antigen; MHC class I antigen A*30; MHC class I antigen A*33; MHC class I antigen A*74; MHC class I antigen B*47; MHC class I antigen B*50; MHC class I antigen B*82; MHC class I antigen Cw*15; MHC class I antigen Cw*18; MHC class I antigen GN00104; MHC class I antigen heavy chain; MHC class I antigen heavy chain HLA-C; MHC class I antigen HLA-A heavy chain; MHC class I antigen HLA-A33; MHC class I antigen HLA-B alpha chain; MHC class I antigen HLA-B heavy chain; MHC class I antigen null protein; MHC class I antigen SHCHA; MHC class I antigene; MHC class I histocompatibility antigen; MHC class I human leukocyte antigen; MHC class I molecule; MHC class I protein; MHC class II antigen; MHC class l antigen; MHC classI antigen; MHC HLA-ABC; MHC HLA-B cell surface glycoprotein; MHC HLA-B transmembrane glycoprotein; MHC HLA-B27d; MHC HLA-B27-HS; MHC HLA-B39N; MHC I HLA; MHC1; PSORS1; SPDA1; surface antigen; truncated class I antigen; truncated MHC class I antigen | |
| IgG2a κ | |
| Affinity chromatography | |
| HLA-B |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title