missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA Class I Antibody (MEM-123), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-505PECY55
This item is not returnable.
View return policy
Description
HLA Class I Monoclonal specifically detects HLA Class I in Human, Bovine, Primate samples. It is validated for Flow Cytometry.
Specifications
| HLA Class I | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| HLA-A | |
| COS-7 African green monkey kidney cells | |
| 0.1 mL | |
| Primary | |
| This antibody (clone MEM-123) reacts with all human classical MHC Class I molecules (major histocompatibility complex) in native cell-surface forms as well as with human HLA-G cDNA transfected cells. MHC Class I molecules (MHC Class Ia) are expressed on the surface of all human cell types. This antibody completely blocks binding of classical W6/32 to surface-expressed HLA-G, but does not cross-blocks This antibody MEM-G/9. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| MEM-123 | |
| Flow Cytometry | |
| A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80 | |
| Mouse | |
| Protein A purified | |
| Adaptive Immunity, Cell Biology, Immunology | |
| 3105.0 | |
| Human, Bovine, Primate | |
| IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction