missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA DOA Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00€ - 470.00€
Specifications
| Antigen | HLA DOA |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18383375
|
Novus Biologicals
NBP3-03621-20UL |
20 μL |
190.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18336224
|
Novus Biologicals
NBP3-03621-100UL |
100 μL |
470.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HLA DOA Polyclonal specifically detects HLA DOA in Human samples. It is validated for Western Blot.Specifications
| HLA DOA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| HLA class II histocompatibility antigen, DO alpha chain, HLA-D0-alpha, HLA-DNAHLADZ, HLA-DZA, lymphocyte antigen, major histocompatibility complex, class II, DN alpha, major histocompatibility complex, class II, DO alpha, MHC class II antigen DOA, MHC DN-alpha, MHC DZ alpha | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 26-135 of human HLA DOA (NP_002110.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Asthma, Diabetes Research, Immunology | |
| PBS with 50% glycerol, pH7.3. | |
| 3111 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title