missing translation for 'onlineSavingsMsg'
Learn More

HLA DQ Antibody (K274.3G8), Novus Biologicals™

Mouse Monoclonal Antibody

188.00€ - 463.00€

Specifications

Antigen HLA DQ
Clone K274.3G8
Dilution Flow Cytometry 1:10 - 1:100, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18678837
View Documents
Novus Biologicals
NBP3-11305-0.1mg
0.1 mg
463.00€
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18666797
View Documents
Novus Biologicals
NBP3-11305-0.025mg
0.025 mg
188.00€
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

HLA DQ Monoclonal antibody specifically detects HLA DQ in Porcine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications

Specifications

HLA DQ
Flow Cytometry 1:10 - 1:100, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Monoclonal
Purified
RUO
Pig
CD, CELIAC1DQ alpha 1 chain, DC-1 alpha chain, DQ-A1, FLJ27088, FLJ27328, GSE, HLA class II histocompatibility antigen, DQ(W3) alpha chain, HLA-DCA, HLA-DQA, leucocyte antigen DQA1, leukocyte antigen alpha chain, major histocompatibility complex, class II, DQ alpha 1, MGC149527, MHC class II antigen, MHC class II DQA1, MHC class II HLA-D alpha glycoprotein, MHC class II HLA-DQ-alpha-1, MHC class II surface glycoprotein, MHC HLA-DQ alpha
Porcine peripheral blood lymphocytes
Primary
Store at 4°C.
K274.3G8
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Unconjugated
Mouse
Adaptive Immunity, Asthma, Immunology
PBS
3117
IgG1
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.