missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA DR/DP Antibody (CL10962) [DyLight 755], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21187IR
This item is not returnable.
View return policy
Description
HLA DR/DP Monoclonal antibody specifically detects HLA DR/DP in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| HLA DR/DP | |
| Monoclonal | |
| DyLight 755 | |
| FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MLRW | |
| This antibody was developed using a synthetic peptide derived from P01903, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Diabetes Research, Immunology | |
| 3122 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL10962 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction