missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EnquireBio HLA-DR Mouse, PE, Clone: L243, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio QAB86-PE-100Tests
Additional Details : Weight : 0.01000kg
Specifications
HLA-DR | |
Monoclonal | |
5μL/Test | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
HLA-DRA | |
Protein A/G Purified | |
RUO | |
3122 | |
IgG2a κ |
Flow Cytometry | |
L243 | |
PE | |
NP_061984 | |
Mouse | |
100 Tests | |
Primary | |
Store at 2-4°C. |