Learn More
HLA DRA Antibody (CL10964) [DyLight 550], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-07996R
Product Code. 30291938
  • 737.00€ / 0.10mL
Description
HLA DRA Monoclonal antibody specifically detects HLA DRA in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HLA DRA
Monoclonal
DyLight 550
FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MLRW
This antibody was developed using a synthetic peptide derived from P01903, with the exact immunogen sequence remaining proprietary.
0.1 mL
Adaptive Immunity, Cell Biology, Diabetes Research, Immunology
3122
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL10964