missing translation for 'onlineSavingsMsg'
Learn More

HLA G Antibody (HLAG/8393R), DyLight 405, Novus Biologicals™

Product Code. 18542518 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18542518 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18542518 Supplier Novus Biologicals Supplier No. NBP320690V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

HLA G Monoclonal antibody specifically detects HLA G in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA G
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HLAG/8393R
Conjugate DyLight 405
Formulation 50 mM Sodium Borate
Gene Alias b2 microglobulin, HLA class I histocompatibility antigen, alpha chain G, HLA class I molecule, HLA G antigen, HLA-6.0, HLAG, HLA-G, HLA-G histocompatibility antigen, class I, G, major histocompatibility complex, class I, G, MHC class I antigen G, MHC class Ib antigen, MHC-G, mutant MHC class I antigen, mutant MHC class Ib antigen, sHLA-G, Soluble HLA class I histocompatibility antigen, alpha chain G
Host Species Rabbit
Immunogen Purified His-tagged HLA G protein.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Autoimmune Diseases, Cancer, Developmental Biology, Immunology, Inflammation, Innate Immunity, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 3135
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.