Learn More
Epredia™ HM 325 Rotary Microtome
Product Code. 15340735 Shop All Epredia Products
Change view
Click to view available options
Includes:
E Blade Holder
ER Blade Holder
N/A
Unit Size:
1 pack
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes unitSize
15340735 E Blade Holder 1 pack
12052999 N/A Each
15348501 ER Blade Holder Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product is under eco-contribution tax.
Product Code. 15340735 Supplier Epredia™ Supplier No. 902100A
 Restricted Item
In Stock
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
Cut high-quality paraffin sections without a power supply with the robust Epredia™ HM 325 Rotary Microtome.
  • One-handed zero orientation head
  • Quick-trim button
  • Section counter
  • Guide-rail guidance for the knife holder
  • Large, easy-to-clean waste tray extends below the blade holder
Ordering Information:
  • Blade holder/Knife holder must be ordered separately unless purchased as a package.
Includes:
  • Standard tools
  • Brushed aluminum cover plate (additional customized cover plates available)
  • Dust cover
  • Instruction manual
  • 902100A also includes a universal cassette clamp (715020S) and a disposable blade carrier E (705800)
  • 902100ER also includes a universal cassette clamp (715020S) and a disposable blade carrier ER (705830)
TRUSTED_SUSTAINABILITY
Specifications
Description HM 325 Rotary Microtome E Package
Product Type Rotary Microtome
Includes E Blade Holder
Depth (English) 19 in.
Depth (Metric) 49 cm
Height (English) 11 in.
Height (Metric) 28 cm
Width (English) 16.5 in.
Width (Metric) 42 cm
Weight (English) 51 lb.
Weight (Metric) 23 kg