missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGB3/HMG4 Rabbit anti-Human, Mouse, Rat, Clone: S01-9C9, Novus Biologicals™
Rabbit Monoclonal Antibody
286.00€ - 581.00€
Specifications
| Antigen | HMGB3/HMG4 |
|---|---|
| Clone | S01-9C9 |
| Dilution | Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunofluorescence 20 μg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18372126
|
Novus Biologicals
NBP3-15041-25UL |
25 μg |
286.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18319853
|
Novus Biologicals
NBP3-15041-100UL |
100 μg |
581.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HMGB3/HMG4 Monoclonal antibody specifically detects HMGB3/HMG4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), ImmunofluorescenceSpecifications
| HMGB3/HMG4 | |
| Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunofluorescence 20 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| High mobility group protein 2a, High mobility group protein 4, high mobility group protein B3, high-mobility group (nonhistone chromosomal) protein 4, high-mobility group box 3, HMG-2a, HMG2AHMG-4, HMG4MGC90319, non-histone chromosomal protein | |
| A synthetic peptide of human HMGB3/HMG4 (Uniprot#: O15347) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| S01-9C9 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Chromatin Research, DNA Repair | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
| 3149 | |
| IgG | |
| Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title