Learn More
HNRNPD Mouse, anti-Human, CoraLite Plus 488, Clone: 2G9E6, Proteintech
Mouse Monoclonal Antibody
145.00€ - 450.00€
Specifications
| Antigen | HNRNPD |
|---|---|
| Clone | 2G9E6 |
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18039645
|
Proteintech
CL488-68236-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18004315
|
Proteintech
CL488-68236-100UL |
100 μL |
450.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The HNRNPD antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HNRNPD. This antibody recognizes human, mouse, rat, pig, rabbit antigen. The HNRNPD antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| HNRNPD | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Mouse | |
| 3184 | |
| IgG2b | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 2G9E6 | |
| Immunofluorescence | |
| Liquid | |
| Human, Mouse, Rat, Pig, Rabbit | |
| AUF1, AUF1A, hnRNP D0, HNRNPD, hnRNPD0, HNRPD, P37 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.