missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNRNPUL2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20143-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to HNRNPUL2.
HNRNPUL2 gene ontology annotations related to this gene include RNA binding.Specifications
| HNRNPUL2 | |
| Polyclonal | |
| Unconjugated | |
| HNRNPUL2 | |
| HNRNPUL2, HNRPUL2, SAF A2, Scaffold attachment factor A2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 221092 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q1KMD3 | |
| HNRNPUL2 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction