Learn More
Hoechst 33258, TRC
Specifications
Specifications
| Chemical Name or Material | 4-[5-(4-Methyl-1-piperazinyl)[2,5'-bi-1H-benzimidazol]-2'-yl]-phenol Trihydrochloride |
| CAS | 23491-45-4 |
| Molecular Formula | C25 H24 N6 O . 3 Cl H |
| Quantity | 500 mg |
| Synonym | Hoechst 33258, p-[5-[5-(4-Methyl-1-piperazinyl)-2-benzimidazolyl]-2-benzimidazolyl]-phenol Trihydrochloride, 2-[2-(4-Hydroxyphenyl)-6-benzimidazolyl]-6-(1-methyl-4-piperazinyl)benzimidazole trihydrochloride, 2-[2-(4-Hydroxyphenyl)-6-benzimidazolyl]-6-(1-methyl-4-piperazyl)-benzimidazole trihydrochloride, 2-[2-(4-Hydroxyphenyl)-6-benzimidazolyl]-6-(1-methyl-4-piperazyl)benzimidazole trichloride, 2-[2-(4-Hydroxyphenyl)-6-benzimidazolyl]-6-(1-methyl-4-piperazyl)benzimidazole-3HCl, 2’-(4-Hydroxyphenyl)-5-(4-methyl-1-piperazinyl)-2,5'-bi-1H-benzimidazole 4-[5-[5-(4-Methyl-1-piperazinyl)-2-benzimidazolyl]-2-benzimidazolyl]phenol trihydrochloride, Bisbenzimide, Bisbenzimide (quenchant), Bisbenzimide trihydrochloride, H 33258 |
| InChI Formula | InChI=1S/C25H24N6O.3ClH/c1-30-10-12-31(13-11-30)18-5-9-21-23(15-18)29-25(27-21)17-4-8-20-22(14-17)28-24(26-20)16-2-6-19(32)7-3-16;;;/h2-9,14-15,32H,10-13H2,1H3,(H,26,28)(H,27,29);3*1H |
| SMILES | Cl.Cl.Cl.CN1CCN(CC1)c2ccc3[nH]c(nc3c2)c4ccc5[nH]c(nc5c4)c6ccc(O)cc6 |
| IUPAC Name | 4-[5-[5-(4-methylpiperazin-1-yl)-1H-benzimidazol-2-yl]-1H-benzimidazol-2-yl]phenol;trihydrochloride |
| Molecular Weight (g/mol) | 533.88 |
| Formula Weight | 532.1312 |