Learn More
Hoefer™ Mini and Standard Electrophoresis Blotting Systems
Product Code. 11935388
Change view
Click to view available options
Includes:
Mini Blot Unit with Semidry Unit
Mini Blot Unit with Transfer Tank
Std Blot Unit with Semidry Unit
Std Blot Unit with Transfer Tank
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
11935388 Mini Blot Unit with Transfer Tank Each
11955388 Std Blot Unit with Transfer Tank Each
11965388 Std Blot Unit with Semidry Unit Each
11945388 Mini Blot Unit with Semidry Unit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Regulatory: This product is under eco-contribution tax.
Product Code. 11935388 Supplier Hoefer™ Supplier No. 326223
  • ONLINE EXCLUSIVE PRICE
    2480.00€
    2057.03€ / Pack of 1
    Save 422.97€
    17% Off
Estimated Shipment: 23-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
Includes
The Mini Electrophoresis Blotting System with Transfer Tank includes the Mighty Small II Mini Vertical Electrophoresis System, Model SE260 Complete; TE22 Mini Tank Blotting Unit; and PS300B Power Supply (300V, 500mA, 90w). The Mini Electrophoresis Blotting System with Semidry Unit includes the Mighty Small II Mini Vertical Electrophoresis System, Model SE260 Complete; Semidry Transfer Unit with Built-In Power Supply, Model TE70XP; and PS300B Power Supply (300V, 500mA, 90w). The Standard Electrophoresis Blotting System with Transfer Tank includes the SE600 Chroma Vertical Electrophoresis System; TE62 Tank Blotting Unit; and PS300B Power Supply (300V, 500mA, 90w). The Standard Electrophoresis Blotting System with Semidry Unit includes the SE600 Chroma Vertical Electrophoresis System; Large Semidry Transfer Unit with Built-In Power Supply, Model TE77XP; and PS300B Power Supply (300V, 500mA, 90w).
Economical equipment packages provide proven electrophoresis solutions for Western blotting
  • Comes with PS300B 300 V, 500 mA, 90 W Power Supply for all units
  • Save time and money with a combination package
Specifications
Content And Storage Mighty Small™ II mini deluxe vertical unit, Mighty Small™ mini transfer tank, power supply
Type Mini Electrophoresis Blotting System
Includes Mini Blot Unit with Transfer Tank
Voltage 300 V
Wattage 90 w
Amperage 500 mA