missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Servalyt™ Carrier Ampholytes

Product Code. 11798028
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11798028 10 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11798028 Supplier Hoefer™ Supplier No. 4294001
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Low molecular-weight molecules of zwitterionic character

Specifications

Chemical Name or Material Carrier Ampholyte
Color Light Brown
For Use With (Application) Used for preparation of tube or slab isoelectric focusing Gels, in free flow or preparative devices, or for rehydration of dried Gels cast on film supports (such as immobilized pH strips or rehydratable slab Gels)
pH 3 to 10
Physical Form Liquid
Quantity 10 mL
Formulation Aqueous solution of amphotheric species comprising Sulfonate, Carboxylic and Amino functions 40% (w/v)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.