Learn More
Hoefer™ SUB13 Midi Submarine Combs
Accessories for SUB13 Midi Submarine
Brand:  Hoefer™ SUB13-C2-16
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12655746
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • Ranges from 10, 12, 16, 20, 24 and 28 Wells
  • 1, 1.5 and 2 mm Thickness
  • 3 to 9.5 mm Well Width
  • 13 min., 85 max. μL Sample Volume
Specifications
Specifications
SUB13 Midi Submarine Gel Electrophoresis Unit
16
0.2 cm
0.21 in.
0.19 in.
Comb
0.07 in.
50 μL
0.55 cm
0.5 cm