missing translation for 'onlineSavingsMsg'
Learn More

Holmium(III) chloride, ultra dry, 99.95% (metals basis)

Product Code. 15457445
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15457445 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15457445 Supplier Thermo Scientific Alfa Aesar Supplier No. 047178.06
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10138-62-2 | Cl3Ho | 271.28 g/mol

It is mainly used in research. It is used in ceramics, lasers, nuclear industries and in paramagnetic. It is also employed in calibration of optical spectrophotometers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10138-62-2
Molecular Formula Cl3Ho
Molecular Weight (g/mol) 271.28
MDL Number MFCD00011051
InChI Key PYOOBRULIYNHJR-UHFFFAOYSA-K
Synonym holmium chloride, holmium trichloride, holmium chloride hocl3, holmium iii chloride, wln: ho n-o3*3, holmium iii chloride, anhydrous, holmium iii chloride, anhydrous, ?hocl3, holmium iii chloride, anhydrous, powder trace metals basis, 3ar,8ar---4,4,8,8-tetrakis 3,5-diethylphenyl tetrahydro-2,2-dimethyl-6-phenyl-1,3-dioxolo 4,5-e dioxaphosphepin
PubChem CID 24992
IUPAC Name holmium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Ho+3]

Specifications

Melting Point 718°C
Physical Form Beads
Quantity 5 g
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 3.70 g/mL
Merck Index 14,4727
Solubility Information Soluble in water.
Formula Weight 271.29
Percent Purity 99.95%
Assay (metals basis)
Chemical Name or Material Holmium(III) chloride, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.