missing translation for 'onlineSavingsMsg'
Learn More

Homopiperazine-N,N’-bis-[2-(ethanesulfonic acid)] Disodium Salt, TRC

Product Code. 30407830
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30407830 25 g 25g
30408524 5 g 5g
30397126 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30407830 Supplier Toronto Research Chemicals Supplier No. TRCH60150025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Homopiperazine-N,N’-bis-[2-(ethanesulfonic acid)] Disodium Salt
Molecular Formula C9 H18 N2 Na2 O6 S2
Quantity 25 g
Synonym Tetrahydro-1H-1,4-diazepine-1,4(5H)-diethanesulfonic Acid Disodium Salt
InChI Formula InChI=1S/C9H20N2O6S2.2Na/c12-18(13,14)8-6-10-2-1-3-11(5-4-10)7-9-19(15,16)17;;/h1-9H2,(H,12,13,14)(H,15,16,17);;/q;2*+1/p-2
SMILES [Na]OS(=O)(=O)CCN1CCCN(CCS(=O)(=O)O[Na])CC1
IUPAC Name 2-[4-(2-sodiooxysulfonylethyl)-1,4-diazepan-1-yl]ethylsulfonyloxysodium
Molecular Weight (g/mol) 360.359
Formula Weight 360.04
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.