Learn More
Brady™ Honeywell Xenon 1950G HD Handheld Scanner
Product Code. 17148654 Shop All Brady Products
Change view
Click to view available options
Connection Type:
USB
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connection Type Unit Size
17148654 USB Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 17148654 Supplier Brady™ Supplier No. 306800
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
The Xenon 1950G HD scanner delivers superior barcode scanning and digital image capture.
  • The scanner incorporates a revolutionary decoding architecture and a custom sensor, enabling extended depth of field, faster reading and improved scanning performance on poor-quality barcodes
  • From high density linear to 2D barcodes found directly on the screen of a mobile device, the 1950G HD scanner decodes virtually all barcodes with ease
Specifications
Description High Density Imager Sensor
Connection Type USB
Power Supply 500 mA (at 5 V DC)
Alternate Part Number XENON 1950GHD