missing translation for 'onlineSavingsMsg'
Learn More

Honokiol, Tocris Bioscience™
GREENER_CHOICE

Product Code. 18094772
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18094772 10 mg 10mg
18025642 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18094772 Supplier Tocris Bioscience Supplier No. 4590/10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Blocks NF-κB activation; also anti-inflammatory and antioxidant

Anti-inflammatory, antioxidant, antitumor and antiangiogenic agent. Blocks NF-κB activation by a variety of stimuli, including TNF and okadaic acid. Shown to scavenge superoxide and peroxyl radicals. Also inhibits a variety of inflammatory responses. Reverses cardiac hypertrophy in mice via SIRT3 activation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35354-74-6
Molecular Formula C18H18O2
Molecular Weight (g/mol) 266.34
InChI Key FVYXIJYOAGAUQK-UHFFFAOYSA-N
Synonym honokiol, 5,3'-diallyl-2,4'-dihydroxybiphenyl, 3,5'-diallyl-4,2'-dihydroxybiphenyl, unii-11513cco0n, chembl16901, 3',5-diallyl-1,1'-biphenyl-2,4'-diol, 4-allyl-2-3-allyl-4-hydroxy-phenyl phenol, 3',5-diallyl-2,4'-biphenyldiol, 3',5-diallylbiphenyl-2,4'-diol, 5,3′-diallyl-2,4′-dihydroxydiphenyl
PubChem CID 72303
ChEBI CHEBI:5759
IUPAC Name 2-(4-hydroxy-3-prop-2-enylphenyl)-4-prop-2-enylphenol
SMILES C=CCC1=CC(=C(C=C1)O)C2=CC(=C(C=C2)O)CC=C

Specifications

Quantity 10 mg
Formula Weight 266.33
Percent Purity >98%
Chemical Name or Material Honokiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.