Learn More
Erlab Horizontal Laminar Flow Hood - Olis 1500
Product Code. 18058211 Shop All Erlab Products
Change view
Click to view available options
Product Type:
Horizontal Laminar Air Flow Cabinet
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
18058211 Horizontal Laminar Air Flow Cabinet Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 18058211 Supplier Erlab Supplier No. NOROLIS19813
  • ONLINE EXCLUSIVE PRICE
    8245.00€
    7674.00€ / Pack of 1
    Save 571.00€
    6% Off
 Restricted Item
Estimated Shipment: 28-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
Efficiency without compromise - Provides superior ISO Class 5 protection and a wide workspace for advanced laboratory applications.
  • Horizontal laminar flow hood Olis
  • Model: 1500
  • Protection provided: ISO Class 5, according to the NF EN ISO 14644 ; HEPA H14 filters - 99.995% MPPS (EN 1822-1:2019 standard)
  • Dimensions: 1595 x 795 x 1075 mm
  • Air flow: 1540 m3/h
  • Voltage/Frequency: 230 V (± 10%) / 50 Hz..
  • Power consumption: 650 W
  • Touch screen
  • Side panels: Transparent P mmA
  • Work surface: Brushed 304 L stainless steel
  • Adjustable base, with or without wheels
  • Electrical outlets: 1 additional electrical outlet
  • Gas, Vacuum tap
  • Standard qualification at installation
  • IQ/OQ qualification at installation
Specifications
Product Type Horizontal Laminar Air Flow Cabinet
Coating Epoxy
Filter Type HEPA
Lighting Type LED
Material Stainless Steel