Learn More
Horst™ Hotbands
Product Code. 11917887
Change view
Click to view available options
Voltage:
77 V
120 V
230 V
Wattage:
26w
62w
66w
70w
80w
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage Wattage Unit Size
11917887 230 V 62w Each
11912033 230 V 66w Each
11927887 120 V 62w Each
11947897 230 V 70w Each
11947877 77 V 26w Each
11982016 230 V 80w Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 11917887 Supplier Horst™ Supplier No. SQ00021
  • ONLINE EXCLUSIVE PRICE
    320.94€
    174.29€ / Pack of 1
    Save 146.65€
    45% Off
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Specifications
Dimensions (L x W) 1.13m
Dimensions (L x W x H) 1.13m
Voltage 230 V
Electrical Requirements 230V Voltage
Wattage 62w
Product Type Hotband