Learn More
Invitrogen™ HOXB5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111191
Description
Immunogen sequence: ASSSHFGAVG ESSRAFPAPA QEPRFRQAAS SCSLSSPESL PCTNGDSHGA KPSASSPSDQ ATSASSSANF TEIDEASASS EPEEAASQLS SPSLARAQPE PMATSTAAPE GQTPQIFPWM R Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 98%.
The Hox proteins are a family of transcription factors that play a role in development and cellular differentiation by regulating downstream target genes. Specifically, the Hox proteins direct DNA-protein and protein-protein interactions that assist in determining the morphologic features associated with the anterior-posterior body axis. Hox proteins are involved in controlling axial patterning, leukemias and hereditary malformations. HoxB5 (homeobox protein Hox-B5), also known as HOX2, HU-1, HOX2A, Hox2.1 or HHO.C10, is a member of the Antp homeobox (Hox) family. It is a 269 amino acid long nuclear protein expressed in the central nervous system. HoxB5 contains one homeobox DNAbinding domain and plays a role in the regulation of lung and gut development, providing cells with positional identities on the anterior-posterior body axis.
Specifications
| HOXB5 | |
| Polyclonal | |
| Unconjugated | |
| HOXB5 | |
| AI385717; HHO.C10; homeo box 2A; homeo box B5; homeobox B5; homeobox protein H24.1; Homeobox protein HHO.C10; homeobox protein Hox-2.1; Homeobox protein Hox-2A; homeobox protein Hox-B5; Homeobox protein Hu-1; homeobox protein Mu-1; HOX2; Hox2.1; Hox-2.1; HOX2A; HOXB5; Hoxb-5; HU-1; RGD1562292 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3215 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P09067 | |
| HOXB5 | |
| Recombinant Protein corresonding to Human HOXB5. Recombinant protein control fragment (Product #RP-108362). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.