missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFR alpha-3/GDNF R alpha-3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99437-100ul
This item is not returnable.
View return policy
Description
GFR alpha-3/GDNF R alpha-3 Polyclonal antibody specifically detects GFR alpha-3/GDNF R alpha-3 in Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| GFR alpha-3/GDNF R alpha-3 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:200-1:1000 | |
| GDNF family receptor alpha 3, GDNF family receptor alpha-3, GDNF receptor alpha-3, GDNFR-alpha-3, GFRa-3, GFR-alpha-3, glial cell line-derived neurotrophic factor receptor alpha-3, GPI-linked receptor | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human GFR alpha-3/GDNF R alpha-3 | |
| 100 μg | |
| Cytokine Research, Immunology | |
| 2676 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction