Learn More
Rotronic™ HP23-AW Set 40 Thermo Hygrometer Set
Product Code. 11995105
Change view
Click to view available options
Type:
Hygrometer Set
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
11995105 Hygrometer Set Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11995105 Supplier Rotronic™ Supplier No. HP23AWSET40FS
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Perfect solution for on-site measurements, this hygrometer is supplied in a tough, lightweight ABS carrying case. Rotronic™ HP23-AW Set 40 Thermo Hygrometer Set includes everything needed for measurement and calibration.
  • Handheld instrument
  • Measurement probe
  • Sample holder
  • Disposable sample container
  • Humidity standards (10%rh, 35%rh, 50%rh, 80%rh)
  • Carrying case
Specifications
Type Hygrometer Set
Battery Type 9V Alkaline
Material ABS Housing
Dimensions (L x W x H) 270 x 70 x 30 mm
Length (Metric) Cable 5 m
Weight (Metric) 198 g
Includes Handheld instrument, probe, sample holder, sample container, humidity standards, carrying case