missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPi2 Antibody (HIC1-2B4.2B) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-18946JF525
This item is not returnable.
View return policy
Description
HPi2 Monoclonal antibody specifically detects HPi2 in Human,Mouse samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Frozen),Immunocytochemistry/ Immunofluorescence
Specifications
| HPi2 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| human pancreatic endocrine islet cell | |
| Human pancreatic enriched islet cells containing low levels of exocrine and ductal cells. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunocytochemistry/Immunofluorescence | |
| HIC1-2B4.2B | |
| 50mM Sodium Borate | |
| Mouse | |
| Tissue culture supernatant | |
| RUO | |
| Human, Mouse | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction