Learn More
STEMCELL Technologies hPSC Genetic Analysis Kit
Codice prodotto. 15851657
Encompass_Preferred
Cambia vista
Click to view available options
Quantity:
1 Kit
Dimensione della confezione:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity Dimensione della confezione
15851657 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 15851657 Fornitore STEMCELL Technologies N. del fornitore 07550
 Restricted Item
Estimated Shipment: 25-11-2026
Aggiungi al carrello
Aggiungi al carrello
missing translation for 'validMainframeMsg'
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
qPCR analysis kit for detecting the majority of karyotypic abnormalities reported in human ES and iPS cells
  • Quickly and cost-effectively detect the 8 most common karyotypic abnormalities reported in human pluripotent stem cells (hPSCs)
  • This qPCR-based kit enables the genetic screening of multiple hPSC lines and contains enough material to analyze 20 individual samples in triplicate
  • It uses double-quenched probes with a 5-carboxyfluorescein (5-FAM) dye to give superior performance over other, single-quenched probes
  • The kit includes qPCR Master Mix, ROX Reference Dye, Buffer, and primer/probe mixes designed to detect the critical minimal regions of the 8 most commonly mutated regions, as well as a primer/probe mix to a control location in the genome
  • A genomic DNA control is also provided with demonstrated normal copy number over the regions of interest
Specifica
Description Pluripotent Stem Cells
Quantity 1 Kit
For Use With (Application) Real Time PCR (qPCR)
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.