missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPSE2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21128-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a heparanase enzyme. The encoded protein is a endoglycosidase that degrades heparin sulfate proteoglycans located on the extracellular matrix and cell surface. This protein may be involved in biological processes involving remodeling of the extracellular matrix including angiogenesis and tumor progression. Alternate splicing results in multiple transcript variants.Specifications
| HPSE2 | |
| Polyclonal | |
| Unconjugated | |
| HPSE2 | |
| heparanase 2, HPA2, HPR2, HPSE2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 368128, 545291, 60495 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| B2RY83, Q8WWQ2 | |
| Hpse2 | |
| HPSE2 Fusion Protein Ag15526 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction