missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HR6B/UBE2B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
415.00€ - 539.00€
Specifications
| Antigen | HR6B/UBE2B |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18214004
|
Novus Biologicals
NBP2-58772 |
100 μL |
539.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18678858
|
Novus Biologicals
NBP2-58772-25ul |
25 μL |
415.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HR6B/UBE2B Polyclonal specifically detects HR6B/UBE2B in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| HR6B/UBE2B | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 7320 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LMRDFKRLQEDPPVGVSGAPSENNIMQWNAVIFGPEGTPFEDGTFKLVIEFSEEYPNKPPTVRFLSKMFHPNVYADGSICLDILQNRWSPTYDVSSILTSIQSLLDEPNPNSPANSQAAQ | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| E2 protein, E2-17kDa, HHR6B, HR6B, RAD6 homolog B, RAD6BEC 6.3.2.19, UBC2, Ubiquitin carrier protein B, ubiquitin-conjugating enzyme E2 B, Ubiquitin-conjugating enzyme E2-17 kDa, ubiquitin-conjugating enzyme E2B (RAD6 homolog), Ubiquitin-protein ligase B | |
| UBE2B | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title