missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HRAS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99921-100ul
This item is not returnable.
View return policy
Description
HRAS Polyclonal antibody specifically detects HRAS in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| HRAS | |
| Polyclonal | |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 4-6 uL/mg of lysate | |
| C-BAS/HAS;C-HA-RAS1;C-H-RAS;CTLO;GTPase HRas;HAMSV;HRAS1;H-RASIDX;p21ras;RASH1 | |
| Produced in rabbits immunized with purified, recombinant Human HRAS (Accession#: P01112; Met1-Cys186) | |
| 100 μg | |
| Cell Cycle and Replication, Metastasis, mTOR Pathway | |
| 3265 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction