Learn More
DNA Antibody (DSD/958), Biotin , Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-11590B
Product Code. 18656077
  • 774.00€ / 0.10mL
Description
DNA Monoclonal antibody specifically detects DNA in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DNA
Monoclonal
Biotin
Double stranded deoxyribonucleic acid, Double stranded DNA, dsDNA
Nuclei of Burkitt's cells
0.1 mL
Primary
Store at 4°C in the dark.
IgG3 κ
Immunohistochemistry, Immunohistochemistry (Paraffin)
DSD/958
PBS
Mouse
Protein G purified