Learn More
Cytiva HRP labelled Protein A
Product Code. 10400325 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10400325 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10400325 Supplier Cytiva Supplier No. NA91201ML
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Protein A conjugated with Horseradish peroxidase for detection of immunoglobulins via affinity to the Fc region - typically IgG from many mammalian species
For use in immunodetection and in situ hybridization applications.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Protein A HRP
For Use With (Application) For detection of immunoglobulins via affinity to the Fc region - typically IgG from many mammalian species
Quantity 1 mL