Learn More
Horst™ HS Heating Wire
Product Code. 10522892
Change view
Click to view available options
Wattage:
100W
250W
350W
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wattage Unit Size
10522892 250W Each
10210402 100W Each
10097151 350W Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 10522892 Supplier Horst™ Supplier No. 20103
  • ONLINE EXCLUSIVE PRICE
    158.01€
    71.82€ / Pack of 1
    Save 86.19€
    54% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Extremely flexible glass yarn-insulated heating cable. Horst™ HS Heating Wire can be used for very small winding radii.
  • Flexible
  • Very small winding radius
Specifications
Length (Metric) 1.5 m
Voltage 230 V
Model HS
Material Glass-yarn
Wattage 250W
Product Type Heating Wire