Learn More
Horst™ HS Model Heating Cable
Product Code. 10057001
Change view
Click to view available options
Model:
HMSG
HS
Length (Metric) Power Cord:
0.6m
1 m
4m
5m
6m
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Length (Metric) Power Cord unitSize
10057001 HS 6m Each
10789442 HMSG 1 m Each
10582502 HS 0.6m Each
10332512 HS 5m Each
10007161 HS 4m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 10057001 Supplier Horst™ Supplier No. 20107
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Specifications
Dimensions (L x W) 6 Lm
Dimensions (L x W x H) 6 Lm
Electrical Requirements ∼230V Voltage
Model HS
Length (Metric) Power Cord 6m
Product Type Heating Cable