Learn More
HSD3B2 Mouse anti-Human, Unconjugated, Clone: 2E7B4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67572-1-IG-150UL
Description
The HSD3B2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HSD3B2. This antibody recognizes human, pig antigen. The HSD3B2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| HSD3B2 | |
| Monoclonal | |
| 900 μg/mL | |
| Mouse | |
| Primary | |
| Human, Pig |
| Western Blot, ELISA | |
| 2E7B4 | |
| Unconjugated | |
| 150 μL | |
| 3284 | |
| IgG1 |