missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSDL1 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00083693-B01P
This item is not returnable.
View return policy
Description
HSDL1 Polyclonal antibody specifically detects HSDL1 in Human samples. It is validated for Western Blot
Specifications
| HSDL1 | |
| Polyclonal | |
| Western Blot 1:500 | |
| NP_113651 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.4) | |
| hydroxysteroid dehydrogenase like 1, inactive hydroxysteroid dehydrogenase-like protein 1, MGC125994, MGC125995, MGC126032, SDR12C3, short chain dehydrogenase/reductase family 12C, member 3, steroid dehydrogenase-like protein | |
| HSDL1 (NP_113651, 1 a.a. - 330 a.a.) full-length human protein. MAAVDSFYLLYREIARSCNCYMEALALVGAWYTARKSITVICDFYSLIRLHFIPRLGSRADLIKQYGRWAVVSGATDGIGKAYAEELASRGLNIILISRNEEKLQVVAKDIADTYKVETDIIVADFSSGREIYLPIREALKDKDVGILVNNVGVFYPYPQYFTQLSEDKLWDIINVNIAAASLMVHVVLPGMVERKKGAIVTISSGSCCKPTPQLAAFSASKAYLDHFSRALQYEYASKGIFVQSLIPFYVATSMTAPSNFLHRCSWLVPSPKVYAHHAVSTLGISKRTTGYWSHSIQFLFAQYMPEWLWVWGANILNRSLRKEALCCTA | |
| 0.05 mg | |
| Lipid and Metabolism | |
| 83693 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction