missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hsp104 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
Brand: Novus Biologicals NBP1-97473-0.025MG
This item is not returnable.
View return policy
Description
Hsp104 Polyclonal specifically detects Hsp104 in Yeast samples. It is validated for Western Blot.
Specifications
| Hsp104 | |
| Polyclonal | |
| Western Blot 1:1000 | |
| Heat shock protein 104, Hsp 104, L0948 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Yeast | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS and 50% Glycerol with 0.09% Sodium Azide | |
| HSP104 | |
| Synthetic peptide corresponding to the sequence near the C-terminus of yeast Hsp104. | |
| 0.025 mg | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction