missing translation for 'onlineSavingsMsg'
Learn More

HSP90 Antibody (2D12), Novus Biologicals™

Rat Monoclonal Antibody has been used in 4 publications

Brand:  Novus Biologicals NBP1-97450-0.2ML

 View more versions of this product

Product Code. 18094159

  • 695.00€ / 0.20mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

HSP90 Monoclonal specifically detects HSP90 in Human, Mouse, Rat, Bovine, Canine, Chicken, Fish, Fungi, Guinea Pig, Hamster, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HSP90
Monoclonal
Unconjugated
PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide
HSP90AA1
Native mouse Hsp90.
0.2 mL
Primary
Reacts with Beluga, and water mold
Store at -20C. Avoid freeze-thaw cycles.
IgM
Western Blot, Immunoprecipitation
2D12
Western Blot 1:500, Immunoprecipitation 1:10-1:500
FLJ31884, Heat shock 86 kDa, heat shock 90kD protein 1, alpha, heat shock 90kD protein 1, alpha-like 4, heat shock 90kD protein, alpha-like 4, heat shock 90kDa protein 1, alpha, heat shock protein 90kDa alpha (cytosolic), class A member 1, heat shock protein HSP 90-alpha, Hsp89, Hsp90, HSP90AHSP86, HSP90N, HSPC1HSP 86, HSPCAHSP89A, HSPCAL1, HSPCAL4, HSPN, LAP2, Renal carcinoma antigen NY-REN-38
Rat
Ammonium sulfate precipitation
RUO
3320
Human, Mouse, Rat, Bovine, Canine, Chicken, Fish, Fungi, Guinea Pig, Hamster, Primate, Rabbit, Sheep
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only