missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HspA6 Antibody (165f), Novus Biologicals™
Mouse Monoclonal Antibody
161.00€ - 769.00€
Specifications
| Antigen | HspA6 |
|---|---|
| Clone | 165f |
| Dilution | Western Blot 1:1000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18074598
|
Novus Biologicals
NBP1-97466-0.025MG |
0.025 mg |
161.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18003009
|
Novus Biologicals
NBP1-97466-0.05MG |
0.05 mg |
391.00€
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18011099
|
Novus Biologicals
NBP1-97466-0.2MG |
0.2 mg |
769.00€
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HspA6 Monoclonal specifically detects HspA6 in Human samples. It is validated for Western Blot.Specifications
| HspA6 | |
| Western Blot 1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
| 3310 | |
| Synthetic peptide corresponding to the sequence near the C-terminus of human HspA6 Antibody (165f). | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| 165f | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| heat shock 70 kDa protein 6, Heat shock 70 kDa protein B', heat shock 70kD protein 6 (HSP70B'), heat shock 70kDa protein 6 (HSP70B'), HSP70B' | |
| HSPA6 | |
| IgG1 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title