missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSPB3 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121893
This item is not returnable.
View return policy
Description
Inhibitor of actin polymerization.
Specifications
| HSPB3 | |
| Polyclonal | |
| Unconjugated | |
| HSPB3 | |
| DHMN2C; heat shock 17 kDa protein; heat shock 27kD protein 3; heat shock 27kD protein family, member 3; heat shock 27kDa protein 3; Heat shock protein; heat shock protein 3; heat shock protein B3; heat shock protein beta-3; heat shock protein family B (small) member 3; HMN2C; HSP; HSP 17; HSP27; HspB3; HSPL27; Protein 3 | |
| Rabbit | |
| Protein A, Antigen affinity chromatography | |
| RUO | |
| 8988 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.66 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q12988 | |
| HSPB3 | |
| Met1-Lys150 (Accession Q12988), with N-terminal His Tag. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction