Learn More
Horst™ HSS Model Flexible Heating Conductor
Product Code. 10532512
Change view
Click to view available options
Dimensions (L x W):
0.6 Lm
1 Lm
10 Lm
2 Lm
3 Lm
4 Lm
5 Lm
6 Lm
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
10532512 4 Lm Each
10280062 0.6 Lm Each
10648852 10 Lm Each
10097071 1 Lm Each
10097081 2 Lm Each
10240112 3 Lm Each
10478261 5 Lm Each
10210212 6 Lm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 10532512 Supplier Horst™ Supplier No. 20905
  • ONLINE EXCLUSIVE PRICE
    178.92€
    116.20€ / Pack of 1
    Save 62.72€
    35% Off
View all promotions
Estimated Shipment: 03-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Specifications
Dimensions (L x W) 4 Lm
Dimensions (L x W x H) 4 Lm
Electrical Requirements ∼230V Voltage
Model HSS
Product Type Flexible Heating Conductor