missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HTR1A Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142775
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
This is one of the several different receptors for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. The activity of this receptor is mediated by G proteins that inhibit adenylate cyclase activity.
Specifications
| HTR1A | |
| Polyclonal | |
| Unconjugated | |
| HTR1A | |
| 5HT1A; 5-HT1A; 5-HT-1A; 5-HT1a receptor; 5-hydroxytryptamine (serotonin) receptor 1A; 5-hydroxytryptamine (serotonin) receptor 1A, G protein-coupled; 5-hydroxytryptamine receptor 1A; ADRB2RL1; ADRBRL1; G-21; Gpcr18; G-protein coupled receptor 18; guanine nucleotide-binding regulatory protein-coupled receptor; HTR1A; PFMCD; RAT5HT1A; Serotonin receptor 1A | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 15550, 3350 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P08908, Q64264 | |
| HTR1A | |
| Peptide with sequence C-ERKNERTAEAKRK (aa 322-334). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction