missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HTR2A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533302
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla (100%); Orangutan, Gibbon, Monkey (95%); Marmoset (85%). Antigen retrieval requires Proteinase K treatment.
In platelets and gut, serotonin plays a major role in cardiovascular function and motility of the gastrointestinal tract, respectively. Serotonin mediates its effects through several of G protein coupled receptors, designated 5-HT receptors or alternatively SR receptors. The SR-2 receptors are comprised of three subtypes, SR-2A, SR-2B and SR-2C, which activate phospholipase C and release intracellular stores of calcium in response to serotonin. SR-2A has a specific role in tracheal smooth muscle contraction, bronchoconstriction and mediating aldosterone production, and it is also thought to play a role in several psychiatric disorders, including depression and schizophrenia. SR-2B is expressed in embryonic and adult cardiovascular tissues, gut and brain and plays an important role in the pathology of cardiac disorders. SR-2C is thought to mediate the effects of atypical antipsychotic drugs.
Specifications
| HTR2A | |
| Polyclonal | |
| Unconjugated | |
| Htr2a | |
| 5-HT-2; 5Ht-2; 5-HT2 receptor; 5-HT2A; 5-HT-2A; 5HT2A Receptor; 5-HT2A receptor; 5-HT2A/2C; 5-HT2A/2C receptor; 5-hydroxytryptamine (serotonin) receptor 2A; 5-hydroxytryptamine (serotonin) receptor 2A, G protein-coupled; 5-hydroxytryptamine 2 receptor; 5-hydroxytryptamine receptor 2A; E030013E04; Htr2; Htr-2; HTR2A; serotonin 5HT-2 receptor; serotonin 5-HT-2A receptor; serotonin receptor 2A; SR2A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3356 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P28223 | |
| Htr2a | |
| Synthetic 20 amino acid peptide from C-terminal cytoplasmic domainhuman 5HT2A Receptor. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction