Learn More
Herka™ Hubbard™ Borosilicate Glass Pycnometer
Product Code. 11900657
Change view
Click to view available options
Capacity (Metric):
30 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11900657 30 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11900657 Supplier Herka™ Supplier No. A1858300
  • ONLINE EXCLUSIVE PRICE
    89.30€
    73.43€ / Pack of 1
    Save 15.87€
    17% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Designed for determining and comparing density or specific gravity values. Herka™ Hubbard™ Borosilicate Glass Pycnometer is suitable for use with liquids or solids.
  • For use with liquids or solids
  • Borosilicate glass construction
Specifications
Capacity (Metric) 30 mL
Material Borosilicate Glass
For Use With (Application) Measuring density and specific gravity