missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ACP6 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7766
This item is not returnable.
View return policy
Description
ACP6 Polyclonal specifically detects ACP6 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| ACP6 | |
| Polyclonal | |
| Unconjugated | |
| BAA91310 | |
| ACP6 | |
| Chinese hamster ovary cell line CHO-derived recombinant human ACP6 Glu33-Glu428 Accession # BAA91310 | |
| 100 μg | |
| Primary | |
| Detects human ACP6 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| acid phosphatase 6, lysophosphatidicPACPL1, Acid phosphatase-like protein 1, ACPL1, ACPL1acid phosphatase like 1, EC 3.1.3.2, LPAP, LPAPlysophosphatidic acid phosphatase type 6, PACPL1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 51205 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction